Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Ethylmorpholine, 98%

Catalog No. p-7081702
Click to view available options
Quantity:
500 mL
1000 mL

CAS: 100-74-3 | C6H13NO | 115.176 g/mol

N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.

Solubility
Miscible with water.

Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-74-3
Molecular Formula C6H13NO
MDL Number MFCD00006177
InChI Key HVCNXQOWACZAFN-UHFFFAOYSA-N
PubChem CID 7525
IUPAC Name 4-ethylmorpholine
SMILES CCN1CCOCC1

Specifications

Melting Point -63°C
Density 0.908
Boiling Point 138°C to 139°C
Flash Point 27°C (80°F)
Refractive Index 1.4413
Quantity 500 mL
UN Number UN2734
Beilstein 102969
Sensitivity Air Sensitive
Synonym n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq
Solubility Information Miscible with water.
Molecular Weight (g/mol) 115.176
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material 4-Ethylmorpholine
Show More Show Less
Hazard Category H226-H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H226
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 202-885-0
RTECSNumber QE4025000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.