Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Ethylmorpholine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1000 mL
Description
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.
Solubility
Miscible with water.
Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.
Solubility
Miscible with water.
Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Melting Point | -63°C |
Density | 0.908 |
Boiling Point | 138°C to 139°C |
Flash Point | 27°C (80°F) |
Refractive Index | 1.4413 |
Quantity | 500 mL |
UN Number | UN2734 |
Beilstein | 102969 |
Sensitivity | Air Sensitive |
Synonym | n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq |
Show More |
Safety and Handling
Hazard Category | H226-H302+H332-H311-H314-H335 |
Hazard Statement | GHS H Statement H301-H311-H314-H318-H226 Toxic if swallowed. Toxic in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S. |
EINECSNumber | 202-885-0 |
RTECSNumber | QE4025000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction