Learn More
4-Fluoro-1,3-dioxolan-2-one, 98%
Description
4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Fluoro-1,3-dioxolan-2-one is used as an electrolyte additive in lithium ion batteries. It also enables the formation of thin, smooth and stable passive solid electrolyte interphase (SEI) layer, which increases the cycling efficiency and discharge capacity retention of the secondary battery. Further, it is used as an intermediate in organic synthesis.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
 
            Specifications
Specifications
| Melting Point | 18°C to 23°C | 
| Color | Colorless | 
| Boiling Point | 212°C | 
| Flash Point | >102°C (216°F) | 
| Quantity | 25 g | 
| Solubility Information | Slightly miscible with water. | 
| Formula Weight | 106.05 | 
| Percent Purity | 98% | 
| Physical Form | Liquid | 
| Chemical Name or Material | 4-Fluoro-1,3-dioxolan-2-one | 
Safety and Handling
| Hazard Category | H302-H315-H319-H335 | 
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. | 
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c | 
| EINECSNumber | 483-360-5 | 
| TSCA | Yes | 
| storageNote1 | 2° to 8°C | 
| Recommended Storage | Keep cold | 
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
 
                                .png-250.jpg) 
                                