Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-N-methylbenzylamine, 97%

Catalog No. p-7036122
Click to view available options
Quantity:
1 g
5 g

CAS: 405-66-3 | C8H10FN | 139.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 405-66-3
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD03839848
InChI Key SZJIQLSCDIEJFC-UHFFFAOYSA-N
Synonym n-methyl-4-fluorobenzylamine, 4-fluoro-n-methylbenzylamine, 4-fluoro-benzyl-methyl-amine, 1-4-fluorophenyl-n-methylmethanamine, n-4-fluorobenzyl-n-methylamine, 4-fluoro-n-methyl benzylamine, 4-fluorophenyl methyl methyl amine, benzenemethanamine, 4-fluoro-n-methyl, 1-4-fluorophenyl-n-methyl-methanamine, acmc-1aek3
PubChem CID 736860
IUPAC Name 1-(4-fluorophenyl)-N-methylmethanamine
SMILES CNCC1=CC=C(C=C1)F

Specifications

Density 1.056
Boiling Point 180°C to 181°C
Flash Point 71°C (160°F)
Refractive Index 1.499
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 139.17
Percent Purity 97%
Chemical Name or Material 4-Fluoro-N-methylbenzylamine
Hazard Category H227-H315-H318-H334-H335
Hazard Statement GHS H Statement
H318-H317-H335-H227-H315
Causes serious eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P285-P302+P352-P304+P340-P305+P351+P338-P310-P332+P313-P342+P311-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.