Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenyl Methyl Sulfone 98.0+%, TCI America™
SDP

Catalog No. F065125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F065125G 25 g
F06515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F065125G Supplier TCI America Supplier No. F065125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 455-15-2
Molecular Formula C7H7FO2S
Molecular Weight (g/mol) 174.189
MDL Number MFCD00039753
InChI Key DPJHZJGAGIWXTD-UHFFFAOYSA-N
Synonym 4-fluorophenyl methyl sulfone, 1-fluoro-4-methylsulfonyl benzene, benzene, 1-fluoro-4-methylsulfonyl, 1-fluoro-4-methylsulphonyl benzene, 1-fluoro-4-methanesulfonylbenzene, 4-fluorophenyl methyl sulphone, 4-fluorophenylmethylsulfone, sulfone, p-fluorophenyl methyl, methyl 4-fluorophenyl sulfone, 1-fluoro-4-methylsulfonyl-benzene
PubChem CID 67994
IUPAC Name 1-fluoro-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)F

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 25 g
Formula Weight 174.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluorophenyl Methyl Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.