Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorophenyl sulfone, 99%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
383-29-9
C12H8F2O2S
254.25
MFCD00000350
PLVUIVUKKJTSDM-UHFFFAOYSA-N
4-fluorophenyl sulfone,bis p-fluorophenyl sulfone,4,4'-sulfonylbis fluorobenzene,bis 4-fluorophenyl sulfone,4,4'-difluorodiphenyl sulfone,4-fluorophenyl sulphone,4-fluorophenylsulfone,benzene, 1,1'-sulfonylbis 4-fluoro,4,4'-difluorodiphenyl sulphone
67842
1-fluoro-4-(4-fluorophenyl)sulfonylbenzene
C1=CC(=CC=C1F)S(=O)(=O)C2=CC=C(C=C2)F
383-29-9
C12H8F2O2S
254.25
MFCD00000350
PLVUIVUKKJTSDM-UHFFFAOYSA-N
4-fluorophenyl sulfone,bis p-fluorophenyl sulfone,4,4'-sulfonylbis fluorobenzene,bis 4-fluorophenyl sulfone,4,4'-difluorodiphenyl sulfone,4-fluorophenyl sulphone,4-fluorophenylsulfone,benzene, 1,1'-sulfonylbis 4-fluoro,4,4'-difluorodiphenyl sulphone
67842
1-fluoro-4-(4-fluorophenyl)sulfonylbenzene
C1=CC(=CC=C1F)S(=O)(=O)C2=CC=C(C=C2)F
Melting Point 97.0°C to 100.0°C
Quantity 100g
Infrared Spectrum Authentic
Color White to Brown
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (FC6H4)2SO2
Formula Weight 254.25
Physical Form Crystals or Powder
Percent Purity 99%
Chemical Name or Material 4-Fluorophenyl sulfone
Show More Show Less