Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methoxybenzylamine hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7149-10-2
C8H11NO2·HCl
189.64
MFCD00012864
LOYPVODLNGWOLM-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzylamine hydrochloride,4-aminomethyl-2-methoxyphenol hydrochloride,vanillylamine hydrochloride,n-vanillylamine hydrochloride,2-vanillylamine hydrochloride,4-hydroxy-3-methoxybenzylamine hcl,phenol, 4-aminomethyl-2-methoxy-, hydrochloride,4-aminomethyl-2-methoxyphenol, chloride,vanillylamine hcl
165576
4-(aminomethyl)-2-methoxyphenol;hydrochloride
COC1=C(C=CC(=C1)CN)O.Cl
7149-10-2
C8H11NO2·HCl
189.64
MFCD00012864
LOYPVODLNGWOLM-UHFFFAOYSA-N
4-hydroxy-3-methoxybenzylamine hydrochloride,4-aminomethyl-2-methoxyphenol hydrochloride,vanillylamine hydrochloride,n-vanillylamine hydrochloride,2-vanillylamine hydrochloride,4-hydroxy-3-methoxybenzylamine hcl,phenol, 4-aminomethyl-2-methoxy-, hydrochloride,4-aminomethyl-2-methoxyphenol, chloride,vanillylamine hcl
165576
4-(aminomethyl)-2-methoxyphenol;hydrochloride
COC1=C(C=CC(=C1)CN)O.Cl
Melting Point 219°C to 221°C
Color White
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HOC6H3(OCH3)CH2NH2·HCl
Beilstein 13, I, 321
Formula Weight 189.64
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Hydroxy-3-methoxybenzylamine hydrochloride
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.