Alfa Aesar™ 4-Hydroxybenzamidine hydrochloride, 95%

Manufacturer: Alfa Aesar™ H3156609

Quantity 10g

 View more versions of this product

Catalog No. AAH3156609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38148-63-9
    Molecular Formula C7H8N2O·HCl
    Formula Weight 172.61
    MDL Number MFCD00136405
    Chemical Name or Material 4-Hydroxybenzamidine hydrochloride

    Specifications

    Formula Weight 172.61
    Molecular Formula C7H8N2O·HCl
    MDL Number MFCD00136405
    CAS 38148-63-9
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number CV6262000
    Melting Point 221° to 224°C
    Quantity 10g
    Chemical Name or Material 4-Hydroxybenzamidine hydrochloride
    Assay 95%

    Description

    Applications
    4-Hydroxybenzamidine hydrochloride is used as a pharmaceutical intermediate and used in organic chemical synthesis.

    Solubility
    Soluble in water.

    Notes
    Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit