Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxymethyl)benzonitrile, 97%

Catalog No. AC454650050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454650050 5 g
AC454650010 1 g
AC454650250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC454650050 Supplier Thermo Scientific Chemicals Supplier No. 454650050
Only null left
Add to Cart

CAS: 874-89-5 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-89-5
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00870633
InChI Key XAASLEJRGFPHEV-UHFFFAOYSA-N
Synonym 4-hydroxymethyl benzonitrile, 4-cyanobenzyl alcohol, p-cyanobenzyl alcohol, ccris 5116, benzonitrile, 4-hydroxymethyl, 4-cyanobenzenemethanol, 4-cyanophenyl methanol, 4-hydroxymethyl-benzonitrile, 4 hydroxymethyl-benzonitrile
PubChem CID 160549
IUPAC Name 4-(hydroxymethyl)benzonitrile
SMILES OCC1=CC=C(C=C1)C#N

Specifications

Melting Point 41.0°C to 42.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 4-(Hydroxymethyl)benzonitrile
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avo

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.