Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 4-IPP
GREENER_CHOICE

Catalog No. 342910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-291-0 10 mg
34-295-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 34-291-0 Supplier Tocris Bioscience™ Supplier No. 3429/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of macrophage migration inhibitory factor (MIF); suicide substrate

Inhibitor of macrophage migration inhibitory factor (MIF). Acts as a suicide substrate to inactivate MIF catalytic and biological functions; inhibits MIF-associated liver enzyme activity in vivo. Also inhibits migration and anchorage independence of human lung adenocarcinoma cell lines in vitro.

Chemical Identifiers

CAS 41270-96-6
Molecular Formula C10H7IN2

Specifications

Quantity 10 mg
Formula Weight 282.08
Purity >99%
Molecular Weight (g/mol) 282.084
Chemical Name or Material 4-IPP
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.