Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diethyl ether, 99+%, extra pure, stabilized with BHT
Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
Glass Bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -116.0°C |
| Acidity | 2mg/100g max. |
| Density | 0.7140g/mL |
| Boiling Point | 34.6°C |
| Quantity | 5 L |
| Flash Point | −45°C |
| Infrared Spectrum | Authentic |
| Packaging | Metal Drum |
| Physical Form | Liquid |
| Refractive Index | 1.3510 to 1.3530 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: May cause drowsiness or dizziness. Harmful if swallowed. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. |
| Precautionary Statement | GHS P Statement: Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge. Store in a well-ventilated place. Keep cool. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction