Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-2-nitroaniline, 97%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
96-96-8
C7H8N2O3
168.15
MFCD00007152
QFMJFXFXQAFGBO-UHFFFAOYSA-N
2-nitro-4-methoxyaniline,4-amino-3-nitroanisole,fast bordeaux gp,2-nitro-p-anisidine,azobase nas,devol bordeaux b,bordeaux gp base,bordeaux gp salt,fast bordeaux gp base,fast bordeaux gp salt
66793
CHEBI:48973
4-methoxy-2-nitroaniline
COC1=CC(=C(C=C1)N)[N+](=O)[O-]
96-96-8
C7H8N2O3
168.15
MFCD00007152
QFMJFXFXQAFGBO-UHFFFAOYSA-N
2-nitro-4-methoxyaniline,4-amino-3-nitroanisole,fast bordeaux gp,2-nitro-p-anisidine,azobase nas,devol bordeaux b,bordeaux gp base,bordeaux gp salt,fast bordeaux gp base,fast bordeaux gp salt
66793
CHEBI:48973
4-methoxy-2-nitroaniline
COC1=CC(=C(C=C1)N)[N+](=O)[O-]
Quantity 25g
Packaging Glass bottle
Melting Point 123.0°C to 126.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3OC6H3(NO2)NH2
Beilstein 13, 521
Solubility Information Solubility in water: slightly soluble
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 4-Methoxy-2-nitroaniline, 98%
Show More Show Less

GHS H Statement:
Fatal in contact with skin.
Fatal if swallowed.
Fatal if inhaled.
Harmful to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-547-2

RTECSNumber : BY4415000

TSCA : TSCA