Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxypyridine N-Oxide 98.0+%, TCI America™
SDP

Catalog No. M240125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M240125G 25 g
M24015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M240125G Supplier TCI America Supplier No. M240125G

Chemical Identifiers

CAS 1122-96-9
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00006207
InChI Key BOFAIBPJCWFJFT-UHFFFAOYSA-N
Synonym 4-methoxypyridine n-oxide, 4-methoxypyridine 1-oxide, 4-methoxypyridine-n-oxide, pyridine, 4-methoxy-, 1-oxide, 4-methoxypyridin-1-ium-1-olate, 4-methoxypyridine-n-oxide hydrate, pubchem2587, acmc-1buuj, 4-methoxypyridinen-oxide, 4-methoxypiridine-n-oxide
PubChem CID 70743
IUPAC Name 4-methoxy-1-oxidopyridin-1-ium
SMILES COC1=CC=[N+](C=C1)[O-]

Specifications

Melting Point 112°C
Quantity 25 g
Formula Weight 125.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxypyridine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.