Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylmorpholine, 99.5%, purified by redistillation, AcroSeal™

Catalog No. AC432800010
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle

CAS: 109-02-4 | C5H11NO | 101.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-02-4
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.15
MDL Number MFCD00006175
InChI Key SJRJJKPEHAURKC-UHFFFAOYSA-N
Synonym n-methylmorpholine, morpholine, 4-methyl, methylmorpholine, 1-methylmorpholine, morpholine, n-methyl, 4-methylmorfolin, 4-methylmorpholin, n-methyl morpholine, 4-methylmorfolin czech, n-methylmorpholin
PubChem CID 7972
IUPAC Name 4-methylmorpholine
SMILES CN1CCOCC1

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -65.0°C
Density 0.9200g/mL
Boiling Point 115.0°C to 116.0°C (750.0 mmHg)
Flash Point 14°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4339 to 1.4359
Quantity 1 L
Merck Index 15,6362
Specific Gravity 0.92
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: miscible with alcohol,ether and benzene
Viscosity 0.91 mPa.s (20°C)
Formula Weight 101.15
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material N-Methylmorpholine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Ground/Bond container and receiving equipment.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water
EINECSNumber 203-640-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.