Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzophenone 97%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Plastic bottle
134-84-9
C14H12O
196.25
MFCD00008553
WXPWZZHELZEVPO-UHFFFAOYSA-N
4-methylbenzophenone,phenyl p-tolyl methanone,4-methylphenyl phenyl methanone,p-methylbenzophenone,methanone, 4-methylphenyl phenyl,p-benzoyltoluene,4-methyl benzophenone,phenyl p-tolyl ketone,benzophenone, 4-methyl,p-benzophenone, methyl
8652
(4-methylphenyl)-phenylmethanone
CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
134-84-9
C14H12O
196.25
MFCD00008553
WXPWZZHELZEVPO-UHFFFAOYSA-N
4-methylbenzophenone,phenyl p-tolyl methanone,4-methylphenyl phenyl methanone,p-methylbenzophenone,methanone, 4-methylphenyl phenyl,p-benzoyltoluene,4-methyl benzophenone,phenyl p-tolyl ketone,benzophenone, 4-methyl,p-benzophenone, methyl
8652
(4-methylphenyl)-phenylmethanone
CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2
Boiling Point 326°C
Flash Point 143°C
Infrared Spectrum Authentic
Melting Point 53°C to 57°C
Color Beige to White
Packaging Plastic bottle
Quantity 50g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3C6H4COC6H5
Beilstein 07, 440
Merck Index 14, 7317
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 196.25
Physical Form Crystalline Powder or Crystals
Percent Purity ≥96.0%
Chemical Name or Material 4-Methylbenzophenone, 97%
Show More Show Less

EINECSNumber : 205-159-1

RTECSNumber : DJ1750000

TSCA : TSCA