Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylmorpholine N-oxide, 50 wt.% solution in water

Catalog No. AC348271000
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle

CAS: 7529-22-8 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylmorpholine N-oxide
Name Note 50 wt.% solution in water
CAS 7529-22-8, 7732-18-5
CAS Min % 50.0, 48.0
CAS Max % 52.0, 50.0
Melting Point -20.0°C
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 118.0°C to 119.0°C
Flash Point >100°C
Packaging Glass bottle
Molecular Formula C5H11NO2
Quantity 100 g
MDL Number MFCD00005947
Fieser 07,244
Specific Gravity 1.13
Merck Index 15, 6171
Synonym 4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f
Solubility Information Solubility in water: miscible
InChI Key LFTLOKWAGJYHHR-UHFFFAOYSA-N
SMILES C[N+]1([O-])CCOCC1
IUPAC Name 4-methyl-4-oxidomorpholin-4-ium
Molecular Weight (g/mol) 117.15
PubChem CID 82029
ChEBI CHEBI:52093
Formula Weight 117.15
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.