Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylsulfonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193847 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Specifications

Chemical Name or Material 4-(Methylsulfonyl)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 149104-88-1
Melting Point 293°C
Color White-Yellow
Molecular Formula C7H9BO4S
Quantity 1 g
MDL Number MFCD01630820
Synonym 4-methylsulfonyl phenylboronic acid, 4-methanesulfonyl phenylboronic acid, 4-methylsulfonyl phenyl boronic acid, 4-methanesulfonyl benzeneboronic acid, 4-methanesulfonylphenylboronic acid, 4-methylsulfonylphenyl boronic acid, 4-boronophenyl methyl sulphone, 4-methylsulphonyl phenylboronic acid, 4-methylsulphonyl benzeneboronic acid, 4-methanesulfonylphenyl boranediol
InChI Key VDUKDQTYMWUSAC-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=C(C=C1)B(O)O
IUPAC Name (4-methanesulfonylphenyl)boronic acid
Molecular Weight (g/mol) 200.02
PubChem CID 2734364
Formula Weight 200.02
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.