Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Morpholinomethyl)aniline, 95+%, Thermo Scientific™

Catalog No. p-4510347
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 51013-67-3
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
InChI Key WNYFVEFUHMDIRQ-UHFFFAOYSA-N
Synonym 4-morpholinomethyl aniline, 4-morpholin-4-ylmethyl aniline, 4-morpholin-4-ylmethylphenylamine, 4-morpholin-4-ylmethyl-phenylamine, benzenamine, 4-4-morpholinylmethyl, 4-4-morpholinylmethyl aniline, 4-morpholin-4-ylmethyl-aniline, 4-morpholin-4-ylmethyl phenylamine, acmc-209krj, 4-morpholinomethyl-aniline
PubChem CID 776851
IUPAC Name 4-(morpholin-4-ylmethyl)aniline
SMILES C1COCCN1CC2=CC=C(C=C2)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 4-(Morpholinomethyl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.