Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Pentylbenzoyl chloride, 98%

Catalog No. AAA1024814
Click to view available options
Quantity:
5 g
25 g
100 g

Catalog No. AAA1024814

Supplier: Thermo Scientific Chemicals A1024814

Only null left
Add to Cart

CAS: 49763-65-7 | C12H15ClO | 210.701 g/mol

4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Pentylbenzoyl chloride was used in the synthesis of 3-O-acyl derivative by reacting with betulinic acid. It is used as intermediates of liquid crystals.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49763-65-7
Molecular Formula C12H15ClO
Molecular Weight (g/mol) 210.701
MDL Number MFCD00000700
InChI Key FBBRKYLXMNQFQU-UHFFFAOYSA-N
Synonym benzoyl chloride, 4-pentyl, p-pentylbenzoyl chloride, 4-n-pentylbenzoyl chloride, 4-n-amylbenzoyl chloride, 4-pentyl-benzoyl chloride, 4-pentylbenzene-1-carbonyl chloride, acmc-20ambq, p-n-amyl benzoyl chloride, 4-n-pentyl benzoyl chloride, 4-n-pentyl-benzoyl chloride
PubChem CID 170812
IUPAC Name 4-pentylbenzoyl chloride
SMILES CCCCCC1=CC=C(C=C1)C(=O)Cl

Specifications

Density 1.034
Boiling Point 144°C to 145°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 25 g
UN Number UN3265
Beilstein 2207887
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 210.71
Percent Purity 98%
Chemical Name or Material 4-n-Pentylbenzoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 256-478-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.