Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridineethanethiol Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. P137325G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. P137325G

Supplier: TCI America P137325G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6298-11-9
Molecular Formula C7H9ClNS
Molecular Weight (g/mol) 174.67
MDL Number MFCD00956047
InChI Key HXMNVHPGTRZNSP-UHFFFAOYSA-N
Synonym 4-(2-Mercaptoethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanethiol Hydrochloride
PubChem CID 11446637
IUPAC Name 2-pyridin-4-ylethanethiol;hydrochloride
SMILES C1=CN=CC=C1CCS.Cl

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 175.67
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyridineethanethiol Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.