Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Thiouridine, 98+%, PROMO, Thermo Scientific Chemicals

Catalog No. J60679MCPM
Click to view available options
Quantity:
100 mg
250 mg

Applications

4-Thiouridine is a constituent of several transfer RNA's. 4-Thiouridine is a thionucleobase used as an antisense agent. Widely used for RNA analysis including RNA-RNA cross coupling and RNA labelling. 4-Thiouridine is a photoreactive (crosslinking) uridine analogue that upon phosphorylation to 4-thioUTP may be incorporated into RNA structures. 4-Thiouridine may be used to study the specificity and kinetics of uridine-cytidine kinases. 4-Thio Uridine is typically used to modify oligos slated for RNA, or RNA-protein, structural studies. A 4-thio-rU modified RNA pentamer was used to study the effect of this modification on codon-anticodon interaction when it is in the wobble position of tRNA.

Solubility

Soluble in water (20 mg/mL), and methanol (10 mg/mL).

Notes

Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13957-31-8
Molecular Formula C9H12N2O5S
Molecular Weight (g/mol) 260.26 g/mol
MDL Number MFCD00006538
InChI Key ZLOIGESWDJYCTF-XVFCMESISA-N
Synonym 4-thiouridine, thiouridine, uridine, 4-thio, 1-beta-d-ribofuranosyl-4-thiouracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-4-thioxo-3,4-dihydropyrimidin-2 1h-one, 1-beta-d-ribofuranosyl-4-thioxo-3,4-dihydropyrimidin-2 1h-one, s4u, thiouridine van, uridine,4-thio, 4-thio-d-uridine
PubChem CID 3032615
ChEBI CHEBI:20480
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(=S)NC1=O

Specifications

Color Yellow
Quantity 100 mg
Solubility Information Soluble in water, and methanol.
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-sulfanylidenepyrimidin-2-one
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material 4-Thiouridine
EINECSNumber 237-735-3
TSCA No
Recommended Storage Store at -20°C

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.