Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)benzyl bromide, 98%, Thermo Scientific™

Catalog No. AC293510100
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 50824-05-0
Molecular Formula C8H6BrF3O
Molecular Weight (g/mol) 255.03
MDL Number MFCD00061238
InChI Key JDNPUJCKXLOHOW-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy benzyl bromide, 1-bromomethyl-4-trifluoromethoxy benzene, 4-trifluoromethoxybenzyl bromide, 4-trifluoromethoxy benzylbromide, p-trifluoromethoxy benzyl bromide, alpha-bromo-4-trifluoromethoxy toluene, benzene, 1-bromomethyl-4-trifluoromethoxy, 4-trifluoromethoxy-benzyl bromide, 4-bromomethyl phenoxy trifluoromethane, 1-bromomethyl-4-trifluoromethoxy-benzene
PubChem CID 142785
IUPAC Name 1-(bromomethyl)-4-(trifluoromethoxy)benzene
SMILES C1=CC(=CC=C1CBr)OC(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 23°C to 24°C
Color Yellow
Density 1.5800g/mL
Boiling Point 82°C to 84°C (10.0 mmHg)
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.482
Linear Formula CF3OC6H4CH2Br
Quantity 10 g
Specific Gravity 1.58
Formula Weight 255.03
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 4-(Trifluoromethoxy)benzyl bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.