Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ High Performance Green Masking Tape

Catalog No. 19070924 Shop All 3M Products
Click to view available options
Quantity:
12/Pk.

Catalog No. 19-070-924

Supplier: 3M™ 7000124898

Only null left
Add to Cart
Add to Cart

3M Company High Performance Green Masking Tape removes cleanly in one piece with no adhesive residue

  • Remains in place around corners, over contours and irregular surfaces
  • Resists paint bleed-through to help create sharp paint lines
  • Green backing improves visibility and accuracy during paint masking
  • Withstands surface temperatures up to 250°F (121°C) for 30 minutes in paint bake operations
  • Deliver sharp paint lines
  • Ideal for critical paint masking applications in automotive, specialty vehicle, and industrial markets, where strong holding and high temp performance is required
  • 48 mm x 55 m 6.7 mil
  • 3M Catalog# 401+

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Material Paper
Product Type Masking and Duct Tapes
For Use With (Application) Painting and Depainting
Model 401+
Quantity 12/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.