Learn More
Description
- Lightweight, unique breathable, open-core padding technology provides optimal airflow and reduces heat and moisture build-up up
- Contoured ergonomic padding design minimizes contact around a worker's neck, lower back and inner thighs to reduce chafing and irritation
- Patented DuraFlex stretchable webbing provides greater flexibility, increased comfort
- Cam buckles provide easy one-hand adjustment
- Holds webbing firmly in place with no readjusting required
- Stand-up back D-ring easily adjusts and attaches
- Quick-connect buckles for easy donning
- Unique and vibrant patterned webbing for easy identification
- Available with quick connect (QC) or tongue buckle, side-D rings, back-D rings
- Recommended Industries/Use: Oil and Gas
- Hazards: Confined Space, fall from height
Certifications
OSHA, ANSI
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI A10.32 and ANSI Z359.1-2007; CSA Z259.10-12 requirements.
Specifications
Specifications
| Includes | Back D-rings, tongue buckle leg, Quick-connect chest |
| D-Ring Location | Back |
| Buckle Style (Leg) | Tongue |
| Capacity (English) | 400 lb. |
| Buckle Style (Chest) | Quick-Connect |
| Capacity (Metric) | 181.5 kg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.