Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ AirCore™ Steel Harnesses

Catalog No. 19053139 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Buckle Style (Leg):
Quick-Connect
Tongue
D-Ring Location:
Back
None
Side
Includes:
Back D-rings, tongue buckle leg, Quick-connect chest
Quick-Connect chest and leg
Side D-rings, Lumbar Pad and Removable Belt Quick-Connect Chest; Tongue Buckle Leg
Side D-rings, Lumbar Pad and Removable Belt, Quick-Connect Chest and Leg
Side D-rings, quick-connect chest and leg
Side D-rings, tongue buckle leg, quick-connect chest
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Buckle Style (Leg) D-Ring Location Includes
19-053-139 Tongue Side Side D-rings, tongue buckle leg, quick-connect chest
19-053-137 Quick-Connect Side Side D-rings, quick-connect chest and leg
19-053-138 Tongue Back Back D-rings, tongue buckle leg, Quick-connect chest
19-053-136 Quick-Connect None Quick-Connect chest and leg
19-066-324 Quick-Connect Side Side D-rings, Lumbar Pad and Removable Belt, Quick-Connect Chest and Leg
19-066-334 Quick-Connect Side Side D-rings, Lumbar Pad and Removable Belt Quick-Connect Chest; Tongue Buckle Leg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-053-139 Supplier Honeywell™ Supplier No. ACTBD/UGN
Add to Cart
Add to Cart

Comes in a high-visibility color for greater safety.

  • Lightweight, unique breathable, open-core padding technology provides optimal airflow and reduces heat and moisture build-up up
  • Contoured ergonomic padding design minimizes contact around a worker's neck, lower back and inner thighs to reduce chafing and irritation
  • Patented DuraFlex stretchable webbing provides greater flexibility, increased comfort
  • Cam buckles provide easy one-hand adjustment
  • Holds webbing firmly in place with no readjusting required
  • Stand-up back D-ring easily adjusts and attaches
  • Quick-connect buckles for easy donning
  • Unique and vibrant patterned webbing for easy identification
  • Available with quick connect (QC) or tongue buckle, side-D rings, back-D rings
  • Recommended Industries/Use: Oil and Gas
  • Hazards: Confined Space, fall from height

Certifications

OSHA, ANSI

Compliance

Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI A10.32 and ANSI Z359.1-2007; CSA Z259.10-12 requirements.

Specifications

Buckle Style (Chest) Quick-Connect
Capacity (Metric) 181.5 kg
Capacity (English) 400 lb.
Buckle Style (Leg) Tongue
D-Ring Location Side
Includes Side D-rings, tongue buckle leg, quick-connect chest
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.