Learn More
Hamilton™ 1000 Series Gastight™ Saltline Syringes
For use with solutions that have a high salt concentration
$95.00 - $201.00
Specifications
Autoclavable Not Autoclavable
Product Type Gastight Saltline Syringe
Connection Type Bubble Free Prime (BFP)
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
14-785-628
View Documents
Hamilton™
208336
5 mL
Each for $125.00
Only null left
Add to Cart
14-785-629
View Documents
Hamilton™
208337
10 mL
Each for $201.00
Only null left
Add to Cart
14-785-627
View Documents
Hamilton™
208335
1 mL
Each for $95.00
Only null left
Add to Cart
Description
Description
  • Equipped with inert and non-corrosive plunger
  • Fitted with resistant ultra-high molecular weight poly-ethylene (UHMWPE) plunger tip
Applications:
For life science applications with salty buffers, mining applications with acids or trace metal analysis
Specifications
Specifications
Not Autoclavable
Gastight Saltline Syringe
Bubble Free Prime (BFP)
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.