Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4EBP1 Antibody (058), Alexa Fluor™ 350
SDP

Catalog No. NB087707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087707 Supplier Novus Biologicals Supplier No. NBP289208AF350

Rabbit Monoclonal Antibody

4EBP1 Monoclonal specifically detects 4EBP1 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen 4EBP1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 58
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias 4E-BP1, eIF4E-binding protein 1, eukaryotic translation initiation factor 4E binding protein 1,4EBP1,4E-BP1BP-1, eukaryotic translation initiation factor 4E-binding protein 1, PHAS-IMGC4316, Phosphorylated heat- and acid-stable protein regulated by insulin 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human 4EBP1 (NP_004086.1; Ser 2-Ile 118).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Lipid and Metabolism, mTOR Pathway, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1978
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.