Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4EBP2 Mouse anti-Human, Clone: 16D9.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS186525UG
Description
Anti-4EBP2, clone 16D9.1 is a mouse monoclonal antibody that detects eIF4E-binding protein 2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Specifications
4EBP2 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
EIF4EBP2 | |
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human eIF4E-binding protein 2. | |
25 μg | |
Signaling | |
NP_004087 | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
16D9.1 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 16D9.1 specifically detects eIF4E-binding protein 2 in human cells. It targets an epitope within 16 amino acids from the C-terminal region. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction