Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4EBP2 Mouse anti-Human, Clone: 16D9.1, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $452.71
Specifications
Antigen | 4EBP2 |
---|---|
Clone | 16D9.1 |
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS1865-25
|
MilliporeSigma
MABS186525UG |
25 μg |
Each for $192.35
|
|
|||||
MABS1865-100
|
MilliporeSigma
MABS1865100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-4EBP2, clone 16D9.1 is a mouse monoclonal antibody that detects eIF4E-binding protein 2 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.Specifications
4EBP2 | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EIF4EBP2 | |
IgG1 κ | |
Protein G Purified | |
Clone 16D9.1 specifically detects eIF4E-binding protein 2 in human cells. It targets an epitope within 16 amino acids from the C-terminal region. |
16D9.1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Signaling | |
NP_004087 | |
KLH-conjugated linear peptide corresponding to 16 amino acids from the C-terminal region of human eIF4E-binding protein 2. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title