Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Diphenylhydantoin, 99%

Catalog No. AAA1701322
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 57-41-0 | C15H12N2O2 | 252.273 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-41-0
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.273
MDL Number MFCD00005264
InChI Key CXOFVDLJLONNDW-UHFFFAOYSA-N
Synonym phenytoin, 5,5-diphenylhydantoin, diphenylhydantoin, dilantin, phenytoine, dihydantoin, aleviatin, dilabid, diphantoin, diphenylan
PubChem CID 1775
ChEBI CHEBI:8107
IUPAC Name 5,5-diphenylimidazolidine-2,4-dione
SMILES C1=CC=C(C=C1)C2(C(=O)NC(=O)N2)C3=CC=CC=C3

Specifications

Melting Point 293°C to 298°C
Quantity 100 g
Beilstein 384532
Merck Index 14,7322
Formula Weight 252.27
Percent Purity 99%
Chemical Name or Material 5,5-Diphenylhydantoin
Hazard Category H302-H350-H361d-H500
Hazard Statement GHS H Statement
H350-H360-H317
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 200-328-6
RTECSNumber MU1050000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.