Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Diaminouracil sulfate, 95%

Catalog No. AC437660500
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

CAS: 42965-55-9 | C4H8N4O6S | 240.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42965-55-9
Molecular Formula C4H8N4O6S
Molecular Weight (g/mol) 240.19
MDL Number MFCD00135845
InChI Key IKARJSDZQCSEJX-UHFFFAOYSA-N
Synonym 5,6-diaminopyrimidine-2,4-diol sulfate, 5,6-diaminopyrimidine-2,4-diol sulphate, diaminopyrimidine-2,4-diol; sulfuric acid, 4,5-diamino-2,6-dihydroxypyrimidine sulfate, 5,6-diaminopyrimidine-2,4 1h,3h-dione sulfate, 5,6-diaminouracil; sulfuric acid, 5?6-diaminouracil sulfate, c4h6n4o2.h2so4, 5,4-dihydroxypyrimidine sulfate, bis 5,6-diaminouracil sulphate
PubChem CID 319643
SMILES OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Brown
Elemental Analysis 18.80 to 21.20% C, 21.93 to 24.73% N
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 50 g
IUPAC Name 5,6-diamino-1,2,3,4-tetrahydropyrimidine-2,4-dione; sulfuric acid
Formula Weight 240.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5, 6-Diaminouracil sulfate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
EINECSNumber 256-022-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.