Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,6-Dihydroxyindole, 95%

Catalog No. AAH6670903
Click to view available options
Quantity:
1 g
5 g

Catalog No. AAH6670903

Supplier: Thermo Scientific Chemicals H6670903

Only null left
Add to Cart

CAS: 3131-52-0 | C8H7NO2 | 149.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3131-52-0
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00798933
InChI Key SGNZYJXNUURYCH-UHFFFAOYSA-N
Synonym 5,6-dihydroxyindole, dopamine lutine, 3h-indole-5,6-diol, 5,6-dihydroxy-1h-indole, dhi, unii-z3oc8499kg, 5,6-dihydroxyindolei, chembl92636, 3id, 5,6-dihydroxy indole
PubChem CID 114683
ChEBI CHEBI:27404
IUPAC Name 1H-indole-5,6-diol
SMILES C1=CNC2=CC(=C(C=C21)O)O

Specifications

Quantity 1 g
Beilstein 122055
Formula Weight 149.15
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 5,6-Dihydroxyindole
Hazard Category H302
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 412-130-9
RTECSNumber NL7858000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.