Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ 5,7-Dichlorokynurenic acid sodium salt

Click to view available options
Quantity:
10 mg
50 mg
Description
5,7-Dichlorokynurenic acid sodium salt is a sodium salt of 5,7-Dichlorokynurenic acid (Cat. No. 0286), a potent antagonist at the glycine site of the NMDA receptor (Ki = 79 nM vs. [3H]-glycine).
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | 5,7-Dichlorokynurenic acid sodium salt |
| Purity | >98% |
| Molecular Weight (g/mol) | 280.04 |
| Inhibitors | 5,7-Dichlorokynurenic acid sodium salt |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction