Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-1H-tetrazole monohydrate, 99%

Catalog No. p-7027343
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 15454-54-3 | CH5N5O | 103.085 g/mol

5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Aminotetrazole monohydrate is a useful reagent in condensation reactions to form pyrimidines.

Solubility
Soluble in water, ethanol (1.36%).

Notes
Store away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15454-54-3
Molecular Formula CH5N5O
Molecular Weight (g/mol) 103.085
MDL Number MFCD00149327
InChI Key JVSMPWHQUPKRNV-UHFFFAOYSA-N
Synonym 1h-tetrazol-5-amine hydrate, 5-amino-1h-tetrazole monohydrate, 5-aminotetrazole monohydrate, 5-amino-1h-tetrazole hydrate, 5-aminotetrazole hydrate, 1h-1,2,3,4-tetrazol-5-amine hydrate, 2h-1,2,3,4-tetrazol-5-amine hydrate, tetrazol-5-amine, 5-aminotetrazole.h2o, 5-aminotetrazole h2o
PubChem CID 12211273
IUPAC Name 2H-tetrazol-5-amine;hydrate
SMILES C1(=NNN=N1)N.O

Specifications

Melting Point ∼203°C (decomposition)
pH 3.7
Quantity 25 g
UN Number UN1325
Beilstein 108872
Solubility Information Soluble in water,ethanol (1.36%).
Formula Weight 103.08 (85.06 Anhydrous)
Percent Purity 99%
Chemical Name or Material 5-Amino-1H-tetrazole monohydrate
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H240-H228-H315-H319-H335
Heating may cause an explosion.
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 224-581-7
RTECSNumber XF7465000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.