Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Amino-2,2-difluoro-1,3-benzodioxole, 97+%

Catalog No. p-7027521
Click to view available options
Quantity:
1 g
5 g

CAS: 1544-85-0 | C7H5F2NO2 | 173.119 g/mol

5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2,2-difluoro-1,3-benzodioxole is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1544-85-0
Molecular Formula C7H5F2NO2
Molecular Weight (g/mol) 173.119
MDL Number MFCD00190144
InChI Key CVYQRDKVWVBOFP-UHFFFAOYSA-N
Synonym 2,2-difluoro-5-aminobenzodioxole, 2,2-difluorobenzo d 1,3 dioxol-5-amine, 2,2-difluoro-5-amino-1,3-benzodioxole, 5-amino-2,2-difluorobenzodioxole, 5-amino-2,2-difluoro-1,3-benzodioxole, 2,2-difluoro-2h-1,3-benzodioxol-5-amine, 1,3-benzodioxol-5-amine, 2,2-difluoro, 3,4-difluoromethylenedioxy aniline, 3,4-difluoromethylene dioxy aniline, 2,2-difluoro-benzo 1,3 dioxol-5-ylamine
PubChem CID 2736893
IUPAC Name 2,2-difluoro-1,3-benzodioxol-5-amine
SMILES C1=CC2=C(C=C1N)OC(O2)(F)F

Specifications

Boiling Point 95°C to 97°C (12 mmHg)
Odor Recognizable
Refractive Index 1.499
Quantity 1 g
UN Number UN2810
Beilstein 1343593
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 173.12
Percent Purity ≥97%
Chemical Name or Material 5-Amino-2,2-difluoro-1,3-benzodioxole
Show More Show Less
Hazard Category H301-H312+H332-H315-H319
Hazard Statement GHS H Statement
H301-H312-H332-H315-H319
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.