Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Azidovaleric Acid 98.0+%, TCI America™
SDP

Catalog No. a2729200mg Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2729200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2729200MG Supplier TCI America Supplier No. A2729200MG
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79583-98-5
Molecular Formula C5H9N3O2
Molecular Weight (g/mol) 143.146
MDL Number MFCD11850108
InChI Key SBZDIRMBQJDCLB-UHFFFAOYSA-N
Synonym 5-Azidopentanoic Acid
PubChem CID 11116171
IUPAC Name 5-azidopentanoic acid
SMILES C(CCN=[N+]=[N-])CC(=O)O

Specifications

Boiling Point 85°C
Quantity 200 mg
Formula Weight 143.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Azidovaleric Acid
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.