Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2,3'-bipyridine, 95%, Thermo Scientific™

Catalog No. p-7030312
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2788003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2788003 Supplier Thermo Scientific Chemicals Supplier No. H2788003
Only null left
Add to Cart

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 774-53-8
Molecular Formula C10H7BrN2
Molecular Weight (g/mol) 235.08
MDL Number MFCD09037420
InChI Key ILJRIAPZWHYPMK-UHFFFAOYSA-N
Synonym 5-bromo-2,3'-bipyridine, 5-bromo-2,3' bipyridinyl, 5-bromo-2-pyridin-3-yl pyridine, 5-bromo-2,3'bipyridine, 5-bromo-2,3-bipyridine, 2,3'-bipyridine, 5-bromo, 5-bromo-2,3' bipyridine
PubChem CID 11390677
SMILES BrC1=CN=C(C=C1)C1=CN=CC=C1

Specifications

Melting Point 75°C to 78°C
Quantity 1 g
IUPAC Name 5-bromo-2,3'-bipyridine
Formula Weight 235.08
Percent Purity 95%
Chemical Name or Material 5-Bromo-2,3'-bipyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.