Learn More
5-Bromo-4-Chloro-3-Indoyl-β-D-Glucuronide sodium salt, 98%, Molecular biology reagent grade, For TLC analysis, MP Biomedicals™
Shop All MP Biomedicals, Inc ProductsSpecifications
Specifications
| Color | White |
| Quantity | 25 mg |
| Formula Weight | 444.6 |
| Grade | Molecular Biology Reagent |
| Physical Form | Powder |
| Chemical Name or Material | 5-Bromo-4-Chloro-3-Indoyl-β-D-Glucuronide sodium salt, For TLC analysis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.