Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorouracil 98.0+%, TCI America™
SDP

Catalog No. C096925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1820-81-1
Molecular Formula C4H3ClN2O2
Molecular Weight (g/mol) 146.53
MDL Number MFCD00006019
InChI Key ZFTBZKVVGZNMJR-UHFFFAOYSA-N
Synonym 5-chlorouracil, 2,4 1h,3h-pyrimidinedione, 5-chloro, uracil, 5-chloro, 5-chloro-2,4-dihydroxypyrimidine, 5-chloropyrimidine-2,4 1h,3h-dione, unii-7lq4v03rny, 7lq4v03rny, 5-chloro-1,3-dihydropyrimidine-2,4-dione, 5-chloropyrimidine-2,4 1h,3h-dione 5-chlorouracil, zlchem 428
PubChem CID 15758
ChEBI CHEBI:60762
IUPAC Name 5-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES ClC1=CNC(=O)NC1=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 146.53
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorouracil
RTECSNumber YQ9410000
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.