Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Fluoro-2-(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.
Solubility
Sparingly Soluble (0.40 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
As raw material in organic synthesis. Utilized in the dye industry. As pharmaceutical intermediate.
Solubility
Sparingly Soluble (0.40 g/L) (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 48°C to 52°C |
| Quantity | 5 g |
| UN Number | UN3261 |
| Synonym | 5-fluoro-2-trifluoromethyl phenol, 5-fluoro-2-trifluoromethyl-phenol, 4-fluoro-2-hydroxybenzotrifluoride, pubchem2171, intermediates-zcf02657, phenol,5-fluoro-2-trifluoromethyl, phenol, 5-fluoro-2-trifluoromethyl |
| Solubility Information | Sparingly Soluble (0.40g/L) (25°C). |
| Formula Weight | 180.1 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Fluoro-2-(trifluoromethyl)phenol |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction