Learn More
5-Formylcytosine (5-fC) Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | 5-Formylcytosine (5-fC) |
| Applications | Dot Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Host Species | Rabbit |
| Immunogen | This 5-Formylcytosine antibody was raised against 5-formylcytidine conjugated to kLH and recognizes 5-formylcytosine. |
| Purification Method | Protein A |
| Quantity | 10 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.