Learn More
5-Hexenyl acetate, 97%
Description
(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.
Solubility
Insoluble in water, soluble in alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Density | 0.883 |
| Boiling Point | 173°C to 174°C |
| Flash Point | 60°C (142°F) |
| Refractive Index | 1.423 |
| Linear Formula | CH3CO2(CH2)4CH=CH2 |
| Quantity | 5 g |
| UN Number | UN3272 |
| Solubility Information | Insoluble in water,soluble in alcohol. |
| Formula Weight | 142.2 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| Hazard Category | H226 |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.