Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Osmium(III) chloride hydrate

Catalog No. AC318700025
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle

CAS: 14996-60-2 | Cl3H2OOs | 314.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14996-60-2
Molecular Formula Cl3H2OOs
Molecular Weight (g/mol) 314.60
MDL Number MFCD00149815
InChI Key KHHOLOMEVBPRQY-UHFFFAOYSA-K
Synonym osmium iii chloride hydrate, osmium chloride oscl3 , hydrate 8ci,9ci, trichloroosmium hydrate, cl3os.h2o, trichloroosmium-water 1/1
PubChem CID 16211485
SMILES O.Cl[Os](Cl)Cl

Specifications

Melting Point >500.0°C
Color Black to Red
Physical Form Crystals or Powder
Quantity 2.5 g
Linear Formula OsCl3·xH2O
Solubility Information Solubility in water: soluble
IUPAC Name trichloroosmium hydrate
Formula Weight 296.56
Packaging Glass bottle
Chemical Name or Material Osmium(III) chloride hydrate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for s

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.