Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-hmC Antibody (AB3/63.3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP25099A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP25099A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP25099A350 Supplier Novus Biologicals Supplier No. NBP250099AF350
Add to Cart
Add to Cart

Rat Monoclonal Antibody

5-hmC Monoclonal specifically detects 5-hmC in All Species samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen 5-hmC
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone AB3/63.3
Conjugate Alexa Fluor 350
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 5′-hmC, 5-hydroxymethylcytosine
Host Species Rat
Immunogen 5-hmC conjugated to BSA
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline DNA Repair, Epigenetics, Stem Cells
Primary or Secondary Primary
Test Specificity Expected to detect 5-hydroxymethylcytosine in all species.
Target Species All species
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.