Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-hmC Antibody (AB3/63.3), PE, Novus Biologicals™
SDP

Catalog No. NB309893 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309893 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309893 Supplier Novus Biologicals Supplier No. NBP250099PE
Add to Cart
Add to Cart

Rat Monoclonal Antibody

5-hmC Monoclonal antibody specifically detects 5-hmC in All samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Dot Blot

Specifications

Antigen 5-hmC
Applications Immunocytochemistry, Immunoprecipitation, Dot Blot
Classification Monoclonal
Clone AB3/63.3
Conjugate PE
Dilution Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Dot Blot
Formulation PBS
Gene Alias 5'-hmC, 5-hydroxymethylcytosine
Host Species Rat
Immunogen 5-hmC conjugated to BSA
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics, Stem Cells
Primary or Secondary Primary
Target Species All species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.