Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-hmC Antibody (C.15200200), Novus Biologicals™
SDP

Catalog No. NBP259177 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259177 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259177 Supplier Novus Biologicals Supplier No. NBP259177
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

5-hmC Monoclonal specifically detects 5-hmC in Human, Mouse samples. It is validated for ELISA, Dot Blot, Methylated DNA Immunoprecipitation.

Specifications

Antigen 5-hmC
Applications ChIP Assay, Dot Blot
Classification Monoclonal
Clone C.15200200
Conjugate Unconjugated
Dilution ELISA 1:500, Dot Blot 2 μg/mL, Methylated DNA Immunoprecipitation 2.5 μg/IP
Formulation PBS with 0.05% Sodium Azide
Gene Alias 5'-hmC, 5-hydroxymethylcytosine
Gene Symbols HMCES
Host Species Mouse
Immunogen The exact immunogen is propietary information.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 56941
Target Species Human, Mouse
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.