Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT1A Antibody, Novus Biologicals™
SDP

Catalog No. NB036666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036666 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036666 Supplier Novus Biologicals Supplier No. NBP289081

Rabbit Polyclonal Antibody

5-HT1A Polyclonal specifically detects 5-HT1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen 5-HT1A
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS containing 50% glycerol
Gene Alias 5-HT-1A, 5-HT1a receptor, 5-HT1A5-hydroxytryptamine receptor 1A, 5-hydroxytryptamine (serotonin) receptor 1A, ADRB2RL15HT1a, ADRBRL1, G protein coupled receptor, G-21, guanine nucleotide-binding regulatory protein-coupled receptor, Serotonin receptor 1A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the sequence near the N-terminus of mouse serotonin receptor 5-HT1A.
Purification Method Peptide affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3350
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.