Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT1A Antibody, Novus Biologicals™
SDP

Catalog No. NBP221590 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP221590 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP221590 Supplier Novus Biologicals Supplier No. NBP221590
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-HT1A Polyclonal specifically detects 5-HT1A in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen 5-HT1A
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Simple Western, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000
Formulation PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal
Gene Accession No. P08908
Gene Alias 5-HT-1A, 5-HT1a receptor, 5-HT1A5-hydroxytryptamine receptor 1A, 5-hydroxytryptamine (serotonin) receptor 1A, ADRB2RL15HT1a, ADRBRL1, G protein coupled receptor, G-21, guanine nucleotide-binding regulatory protein-coupled receptor, Serotonin receptor 1A
Gene Symbols HTR1A
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human 5-HT1A receptor. The exact sequence is proprietary.
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3350
Test Specificity Human, Mouse, Rat. Expected to cross react based on sequence identity: Rabbit (92%), Rhesus Monkey (100%), Chimpanzee (100%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.