Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human 5-HT1B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5858
This item is not returnable.
View return policy
Description
5-HT1B Monoclonal specifically detects 5-HT1B in Human samples. It is validated for Immunohistochemistry.Specifications
| 5-HT1B | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P28222 | |
| HTR1B | |
| NS0 mouse myeloma cell line transfected with human 5HT1B Met1-Ser390 Accession # P28222 | |
| 100 μg | |
| Primary | |
| Detects human 5HT1B. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 499325 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5HT1B, 5-HT1B5-HT-1B, 5HT1DB, 5-HT1DB, 5-hydroxytryptamine (serotonin) receptor 1B, HTR1B, HTR1D2, HTR1D2,5-hydroxytryptamine receptor 1B, HTR1DB, S12, S125-HT-1D-beta, Serotonin 1D beta receptor, Serotonin receptor 1B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3351 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction