Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT1E Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB034550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB034550 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB034550 Supplier Novus Biologicals Supplier No. NBP286928

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

5-HT1E Polyclonal specifically detects 5-HT1E in Human samples. It is validated for Western Blot.

Specifications

Antigen 5-HT1E
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose
Gene Alias 5-HT-1E, 5-hydroxytryptamine (serotonin) receptor 1E, S31,5-HT1E5-hydroxytryptamine receptor 1E, Serotonin receptor 1E
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of Human 5-HT1E
Quantity 100 μg
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 3354
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.